Learn More
Trajan Scientific and Medical™ Replacement Needle and Plunger Kit for Shimadzu Autosampler Syringes
Product Code. 10712772
Click to view available options
Needle Gauge:
23 G
26 G
Diameter (Metric) Outer:
0.47 mm
0.63 mm
Unit Size:
Each
Product Code. 10712772
Brand: Trajan Scientific and Medical™ 033745
Repair kit for Shimadzu Removable Needle Autosampler Syringes.
For use with Shimadzu Removable Needle Syringes for AOC14, AOC17 and AOC 20 Autosamplers
  • Includes spare needle and plunger
  • Plunger extends to needle tip
  • Sample contained in needle only
  • Zero dead volume
  • Needle and plunger must be replaced together
Specifications
Needle Gauge 23 G
Diameter (Metric) Outer 0.63 mm
Product Type Replacement Needle Kit
Needle Point Style Cone
For Use With (Equipment) Syringes (0.5μL)