Learn More
SCAT Replacement Lid for Funnel MARCO V2.0
Product Code. 17383761 Shop All SCAT Europe Products
Change view
Click to view available options
For Use With (Equipment):
Funnel MARCO V2.0
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17383761 Funnel MARCO V2.0 Each
1 options
Product Code. 17383761
Brand: SCAT 318998
Only null left
Add to basket
Add to basket
Replacement lid for funnel MARCO, V2.0, white, not conductive
  • Replacement lid
  • For funnel MARCO, V2.0
  • White
  • Not conductive
Specifications
Material HDPE-EC
Height (Metric) 20 mm
Color Black
For Use With (Equipment) Funnel MARCO V2.0
Diameter (English)Top 140 mm
Product Type Replacement Lid