Learn More
Cole-Parmer™ Replacement Items for Freezer/Mill™ Grinding Vial Sets
Shop All SPEX Certiprep Products
Click to view available options
Product Type:
Center Cylinder
End Plug
Replacement Impactor
Material:
Polycarbonate
Stainless Steel (440 C)
For Use With (Equipment):
6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set
6881 Vial Set
Spex SamplePrep Freezer/Mill Cryogenic Grinder
Unit Size:
Each
Pack of 2
Pack of 20
Pack of 4
Description
- Suitable for use with the Cole-Parmer™ Freezer/Mill™ grinding vial sets
- Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinding vial sets with a wide range of grinding ball options suitable for your application.
Specifications
Specifications
| Product Type | Center Cylinder |
| Disposable | Single-use |
| For Use With (Equipment) | Spex SamplePrep Freezer/Mill Cryogenic Grinder |
| Material | Polycarbonate |
| Model | Cole-Parmer Freezer/Mill |
| Packaging Quantity | 20 Pk. |
| Sample Volume (English) | 0.01 to 0.13 oz. |
| Sample Volume (Metric) | 0.5 to 4 mL |