Learn More
Wheaton Science Products Replacement Glass Impeller for MAGNAFLEX 125ml
Brand:  Wheaton Science Products 356841
Replacement Glass Impeller for MAGNAFLEX 125ml
Product Code. 10681212
  • ONLINE EXCLUSIVE PRICE
    2895.00 NOK
    1966.11 NOK / Each
    Save 928.89 NOK
    32% Off
Only null left
Estimated Shipment: 29-05-2026