Learn More
Idex Replacement Ferrules for SealTight™ Fittings
Product Code. 11588670 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1/Ea
10/pk
Unit Size:
Each
Pack of 10
Product Code. Quantity unitSize
11588670 10/pk Pack of 10
11593970 1/Ea Each
2 options
Product Code. 11588670
Brand: Idex F192X
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Slotted end on ferrule creates second compression zone for a tight seal
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
Color Black
Description 10-32 or M6 coned, for 1/16 in. OD tubing
For Use With (Equipment) 0.0625 in. O.D. Tubing
Material PEEK
Quantity 10/pk