Learn More
UVP Replacement Bulbs
Product Code. 12863848
Change view
Click to view available options
For Use With (Equipment):
B-100 High Intensity UV Lamp
Models C-70G, C-71 Chromato-vue Cabinet
Models UVGL-48, ML-49, UVG-47 UV Lamp
Wattage:
6 W
25 W
100 W
Unit Size:
Each
Product Code. For Use With (Equipment) Wattage unitSize
12863848 Models C-70G, C-71 Chromato-vue Cabinet 25 W Each
12822969 Models UVGL-48, ML-49, UVG-47 UV Lamp 6 W Each
11890483 B-100 High Intensity UV Lamp 100 W Each
3 options
Product Code. 12863848
Brand: UVP 34000201
Specifications
Voltage 115 V
Wattage 25 W
For Use With (Equipment) Models C-70G, C-71 Chromato-vue Cabinet
Includes Replacement 25 W tube/bulb
Packaging Quantity 1 Ea.
Product Type Replacement Bulb