Learn More
UVP Replacement Bulbs
Product Code. 12822969
Change view
Click to view available options
For Use With (Equipment):
B-100 High Intensity UV Lamp
Models C-70G, C-71 Chromato-vue Cabinet
Models UVGL-48, ML-49, UVG-47 UV Lamp
Wattage:
6 W
25 W
100 W
Unit Size:
Each
Product Code. For Use With (Equipment) Wattage unitSize
12822969 Models UVGL-48, ML-49, UVG-47 UV Lamp 6 W Each
11890483 B-100 High Intensity UV Lamp 100 W Each
12863848 Models C-70G, C-71 Chromato-vue Cabinet 25 W Each
3 options
Product Code. 12822969
Brand: UVP 34001401
Only null left
Specifications
Voltage 115 V
Wattage 6 W
Wavelength 365/254 nm
Dimensions (L x W) 9.5 x 2.8 in. (24.1 x 7.1 cm)
For Use With (Equipment) Models UVGL-48, ML-49, UVG-47 UV Lamp
Includes Replacement 6 W tube
Length (English) 9.5 in.
Length (Metric) 24.1 cm
Packaging Quantity 1 Ea.
Width (English) 2.8 in.
Width (Metric) 7.1 cm
Product Type Replacement Bulb