Learn More
UVP Replacement Bulbs
Click to view available options
For Use With (Equipment):
B-100 High Intensity UV Lamp
Models C-70G, C-71 Chromato-vue Cabinet
Models UVGL-48, ML-49, UVG-47 UV Lamp
Wattage:
6 W
25 W
100 W
Unit Size:
Each
Specifications
Specifications
| Base Style | Ad-medium |
| Lighting Life | 16,000 hr. |
| Wattage | 100 W |
| Wavelength | 365 nm |
| For Use With (Equipment) | B-100 High Intensity UV Lamp |
| Includes | Replacement 100 W bulb |
| Length (English) | 7.5 in. |
| Length (Metric) | 19.05 cm |
| Packaging Quantity | 1 Ea. |
| Shape | PAR38 |
| Show More |