Learn More
Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths
Description
- Stainless steel construction
- Works in conjunction with lid to create a drip collection unit
- Prevents user from coming into contact with chemical solutions
- Prevents items from resting on the inside of the tank wall
- For use with Grant Ultrasonic Baths
Specifications
Specifications
| For Use With (Equipment) | XUB5 Ultrasonic Bath |
| Material | Stainless Steel |
| Size | (WxDxH) 265 x 120 x 140 mm |