Description
- Includes 1 allen key, 1 piston rod with fitted PTFE seal, 1 calibrating gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 PTFE seals
- Mounting block included for capacities of 20μL and up
Specifications
Specifications
| Certifications/Compliance | DE-M marked |
| Volume (Metric) | 100 and 200 μL |
| For Use With (Equipment) | Transferpettor™ |
| Includes | Blue mounting block |
| Quantity | 1 Each |