Learn More
Thermo Scientific™ Repair Kits for Thermo Scientific™ Plunger-in-Needle Syringes
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Repair Kits for Thermo Scientific Plunger-in-Needle Syringes extend the life and performance and reduce cost per use of syringes.
- Repair kits for plunger in needle syringes contain both the plunger and needle, which must be changed together
Includes: Plunger, 1; Needle, 1
Specifications
Specifications
| Description | For PIN syringe 365B0531 |
| Needle Gauge | 23 G |
| Length (English) Needle | 2.7559 in. |
| Product Type | Syringe Repair Kit |
| Volume (Metric) | 1 μL |
| Length (Metric) Needle | 70 mm |
| For Use With (Equipment) | PIN Syringe 365B0531 |
| Connection Type | Plunger-In |