Learn More
Corning™ CoolCell™ FTS30 Vial Module
BIOPHARMA
Product Code. 15515535
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15515535 White Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15515535 Supplier Corning™ Supplier No. 432078
  • ONLINE EXCLUSIVE PRICE
    1540.00 NOK
    1457.17 NOK / Pack of 10
    Save 82.83 NOK
    5% Off
Estimated Shipment: 11-03-2027
For use with CoolCell™ FTS30. Corning CoolCell™ FTS30 Vial Module is a cryogenic vials holder that allows one-step insertion and removal of all 30 vials at once.
Corning CoolCell FTS30 Vial Module is a holder for 30 1.0 mL or 2.0 mL cryogenic vials that allows one-step insertion and removal of all 30 vials at once. It fits into a standard 5.0 x 5.0 x 2.0 inch cryostorage box and is compatible with dry ice and liquid nitrogen.
TRUSTED_SUSTAINABILITY
Specifications
Color White
For Use With (Application) Holding cryogenic vials
No. per Case 10
No. per Pack 10