Learn More
GA International Removable Cryogenic Labels for Laser Printers
Product Code. 17814376
Encompass_Preferred
Change view
Click to view available options
Dimensions (LxW):
24 x 13 mm
31 x 22 mm
36 x 14 mm
45 x 20 mm
63.5 x 25.4 mm
Unit Size:
Pack of 1152
Pack of 1600
Pack of 2560
Pack of 528
Pack of 896
Product Code. Dimensions (LxW) unitSize
17814376 24 x 13 mm Pack of 2560
17864786 63.5 x 25.4 mm Pack of 528
17844416 36 x 14 mm Pack of 1600
17804896 45 x 20 mm Pack of 896
17814216 31 x 22 mm Pack of 1152
5 options
Product Code. 17814376
Brand: GA International A4RCL12T1WH
Only null left
Add to basket
Add to basket
Removable cryo labels provided in A4 sheet format for printing with laser printers.
  • Glove-friendly adhesive ensures the labels can be removed easily without leaving any glue residue
  • Resist extreme cryogenic temperatures in liquid and vapor phase liquid nitrogen, up to -196°C
  • Printable by all major desktop laser printers
  • Resistant to gamma irradiation (tested up to 50 KGy)
Specifications
Adhesive Type Removable
Color White
Finish Medium Gloss
Format Sheet
Material Thermoplastic Film
Print Technology Laser
Product Type Label
Sterility Non-sterile
Labels per Sheet 160/Sheet
No. per Pack 16/Pk.
Quantity 2560/Pk.
Shape Rectangle
Temperature Range (English) -321°F to +248°F
Temperature Range (Metric) -196°C to +120°C
Dimensions (LxW) 24 x 13 mm
Length (English) 0.51 in.
Length (Metric) 13 mm
Width (English) 0.95 in.
Width (Metric) 24 mm