Learn More
PureAire Monitoring Systems, Inc. Remote Digital LED Display for Monitors
Product Code. 16268821
Change view
Click to view available options
Length (English):
2 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
16268821 2 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16268821 Supplier PureAire Monitoring Systems, Inc. Supplier No. 99091
  • ONLINE EXCLUSIVE PRICE
    6730.00 NOK
    4863.22 NOK / Pack of 1
    Save 1866.78 NOK
    27% Off
Estimated Shipment: 11-02-2027
The Remote Digital Display provides continuous information on the status of the PureAire Oxygen Deficiency Monitor by remotely mirroring the monitor’s display and alarm panel. A large, bright red LED display continuously illuminates the O2 reading from the oxygen monitor up to 75 meters.
  • Bright 1 in. red LED digital concentration display
  • Built-in alarm and fault status LED’s
  • User selectable dual level alarm set points
  • Built-in audio horn alerts personnel to low or high oxygen conditions
  • Two tone horn distinguishes between alarm 1 and alarm 2
  • Displays Oxygen monitor overall status
  • The Remote Digital Alarm Indicator provides continuous information on the status of the PureAire oxygen deficiency monitors or analysers by remotely mirroring the monitor’s display. A large, bright red LED display continuously illuminates the O2 reading from the oxygen monitor up to 250 feet away.
  • You can select two individual alarm set points to activate the visual alarm LED’s and audible horn. High intensity LED’s illuminate whenever preset alarm levels are exceeded. Also included is an additional fault LED indicator for system errors. The remote digital alarm indicator has a 90dB built-in horn to alert personnel when an unsafe condition exists.
  • It connects to an oxygen monitor using a 3-wire, 18 gauge shielded cable. The remote display can also be connected to a Programmable Logic Controller (PLC), providing simultaneous alarm and status indications in multiple locations throughout the facility.
Warranty
2 years
Specifications
Description Horn Sound Output 80 – 85 db
Dimensions (L x W x H) 2 x 4.6 x 3.5 in.
Height (English) 3.5 in.
Height (Metric) 88.9 mm
Length (English) 2 in.
Length (Metric) 50.8 mm
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40 to +50°C
Voltage 24 VDC
Warranty 2 Year after ETD
Weight (English) 1.1 lb.
Weight (Metric) 0.5 kg
Width (English) 4.6 in.
Width (Metric) 116.84 mm