Learn More
Thermo Scientific™ T Haemagglutination Test
Product Code. 13235789
Change view
Click to view available options
Quantity:
50 Tests/Kit
Unit Size:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13235789 50 Tests/Kit 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13235789 Supplier Thermo Scientific™ Supplier No. R30850601
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Semi-quantitatively measure auto-antibodies to human thyroglobulin using T Haemagglutination Test.
Semi-quantitatively measure auto-antibodies to human thyroglobulin (thyroid auto-antibodies) using Thermo Scientific™ Remel™ Thymune™—T Haemagglutination Test. This test is based upon Boyden's passive haemagglutination system, first used for detection of thyroglobulin antibodies by Witebsky and Rose8. it has been established that auto-antibodies to several different thyroid constituents1,4 are associated with destructive inflammatory lesions of the thyroid gland. Antibodies to these constituents, namely thyroglobulin and the microsomal antigen, are of particular importance for diagnostics purposes3,5,7.
It has been established that autoantibodies to several different thyroid constituents are associated with destructive inflammatory lesions of the thyroid gland. Thyroglobulin and microsomal haemagglutination tests give useful predictive evidence of possible thyroid dysfunction in patients with other autoimmune endocrine disorders such as Addison’s disease, insulin-dependent diabetes mellitus or polyendocrine autoimmunopathies, and in members of families prone to organ-specific autoimmunity. The antibodies detected by the microsomal haemagglutination test are the principal circulating marker of human autoimmune thyroid disease, which include the clinical disorders of goitrous thyroiditis (Hashimoto’s disease), atrophic thyroiditis (myxoedema) and thyrotoxicosis (Graves’/Basedow’s disease)3.
  • A simple and reproducible test
  • Easy to read: Cells coated with thyroglobulin are agglutinated by specific autoantibodies, yielding an even carpet of cells at the bottom of a microtitre well and indicating a positive reaction; lack of agglutination is indicated by the cells settling into a tight ring or button, indicating a negative reaction.
  • The combination of thyroglobulin and microsomal haemagglutination tests will detect practically all Hashimoto goitres and about 90% of primary myxoedema cases.
  • Differential Diagnosis of primary thyrotoxicosis and various tachycardias, anxiety states, unexplained weight loss or diarrhoea.
Proteins such as thyroglobulin are readily bound to the surface of red blood cells which have been treated with tannic acid. Thyroglobulin extracted from human thyroid glands by classical salt precipitation techniques coupled to tanned turkey red cells provides a haemagglutination test system for the detection of low levels of auto-antibody to thyroglobulin. A small proportion of human sera are reactive against turkey cells, giving rise to non-specific agglutination of sensitised cells. These non-specific reactions may be detected by means of unsensitised control cells. Both test and control cells are treated with formalin and freeze dried to give long term stability on storage.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description Thymune-T
CE Marker Yes
Quantity 50 Tests/Kit