Learn More
Thermo Scientific™ Remel™ Sabouraud Dextrose Agar, pH 5.6
Product Code. p-4524398
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. 12677619
Brand: Thermo Scientific™ R454462
Remel Sabouraud Dextrose Agar, pH 5.6 is used for the isolation of fungi.
Remel Sabouraud Dextrose Agar, pH 5.6 is used for the isolation of fungi.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: USP
Description Sabouraud Dextrose Agar, pH 5.6
Form Powder
Product Type Agar
Quantity 500 g
Yield For 7.7L medium
For Laboratory Use Only