missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Phosphate Buffer w/MgCl 99mL
Description
Thermo Scientific™ Remel Phosphate Buffer w/MgCl 99mL (pH 7.2 stock solution) formulation is used in water and wastewater testing as referenced in APHA and EPA test methods.
- pH 7.2 stock solution
- Formulation is used in water and wastewater testing as referenced in APHA and EPA test methods
- Red cap
Specifications
Specifications
| Description | Phosphate Buffer w/MgCl 99mL |
| Format | Red Cap - 99mL/Bottle |
| Product Type | Buffer Solution |
| Packaging Type | 72/Case |
| Quantity | 72 x 99 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?