Learn More
Thermo Scientific™ PathoDx™ Respiratory Virus Panel Influenza B Reagent
Product Code. 11444129
Change view
Click to view available options
Quantity:
1 x 10 mL
Unit Size:
40 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444129 1 x 10 mL 40 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11444129 Supplier Thermo Scientific™ Supplier No. R62406
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PathoDx™ Respiratory Virus Panel Influenza B Reagent along with RVP kit is a direct immunofluorescence test used to detect Influenza B.
Use Thermo Scientific™ Remel™ PathoDx™ Respiratory Virus Panel Influenza B Reagent along with PathoDx Respiratory Virus Panel (RVP) kit to perform a direct immunofluorescence test for the qualitative detection of Influenza B in prepared patient specimens following growth in cell culture. Influenza viruses are negative-stranded RNA viruses of the Orthomyxoviridae family. Influenza infections are associated with seasonal outbreaks of respiratory disease, such as pharyngitis, croup, bronchitis, and pneumonia1. Usually one type, either type A or B, will predominate during each epidemic2.
Influenza B is comprised of a single genus and shows antigenic variation in the hemagglutinin and neuraminidase proteins.
  • Ready to use – Prepared reagents
  • Easy to store – Store at 2-8°C
  • Easy to interpret – Fluorescence is nuclear, cytoplasmic, or both. Nuclear staining is uniformly bright, and cytoplasmic staining is often punctated with large inclusions
Influenzae B Reagent (Purple cap) is one dropper bottle containing 2.0 mL (approximately 40 tests) of fluorescein-labeled purified specific murine monoclonal antibodies, and Evans blue counterstain in a protein stabilized buffer solution, with 0.098% sodium azide as a preservative.
Influenza B can be isolated in MRC-5, RD, A549, HL, and MDCK tube or shell vial cultures with or without the presence of trypsin3,1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Remel™ PathoDX™ Respiratory Virus Panel Influenza B Reagent
Detectable Analytes Influenza
Sample Type Nasopharyngeal
Type Reagent
Quantity 1 x 10 mL