Learn More
Thermo Scientific™ LAP Disk w/Reagent
Product Code. 13205789
Click to view available options
Quantity:
50 Tests/Kit
Unit Size:
50 tests
Product Code. 13205789
Brand: Thermo Scientific™ R30168501
Rapidly detect leucine aminopeptidase activity in streptococci and related genera using LAP Disk with Reagent.
Rapidly detect leucine aminopeptidase activity in streptococci and related genera using Thermo Scientific™ Remel™ LAP Disk with Reagent. In 1984, Colman and Ball reported that, unlike streptococci, Aerococcus species do not produce leucine aminopeptidase (LAP)1. Aerococcus strains were found to be consistently negative for leucine aminopeptidase, while strains of Enterococcus and Streptococcus gave positive reactions. LAP, in conjunction with PYR and other conventional tests, is an important parameter in the identification of Aerococcus, Leuconostoc, Pediococcus, and related genera2-4.
Leucine aminopeptidases (LAPs) are a diverse set of exopeptidases that catalyze the hydrolysis of leucine residues from the amino termini of proteins or peptides. LAPs are ubiquitous enzymes present among animals, plants, prokaryotes and microbes. The LAP test should be performed on pure or isolated cultures of catalase-negative, Gram-positive cocci which are 18-72 hours old.
  • Ready to use: Reagent-impregnated disc.
  • Rapid test: Results are obtained within 5 to 6 minutes.
  • Easy to identify: A color change from pink to red indicates a positive test whereas no color change or a slightly yellow color indicates a negative test.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description Murex Lap Kit/50/Pk.
CE Marker Yes
Includes LAP disk and color developer included
Quantity 50 Tests/Kit
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.