Learn More
Thermo Scientific™ Remel™ Gram Safranin
Código de producto. 12623668
missing translation for 'orderingAttributeHoverText'
Quantity:
5/Pk.
Each
missing translation for 'unitSize'
250mL
4 L
Pack of 5
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 12623668
missing translation for 'mfr': Thermo Scientific™ R40059
Este artículo no se puede devolver. Vea la política de devoluciones
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
TRUSTED_SUSTAINABILITY
Especificaciones
Type Safranin
Quantity 5/Pk.
Certifications/Compliance CE Marked
Volume (Metric) 5 x 250 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.