Learn More
Thermo Scientific™ Remel™ Egg Yolk Tellurite 100%
Click to view available options
Quantity:
100 mL
Packaging Type:
1 Ea.
Unit Size:
100mL
Description
Remel Egg Yolk Tellurite 100% is used with Baird Parker Agar Base.
Specifications
Specifications
| Description | Egg Yolk Tellurite 100% |
| Form | Liquid |
| Product Type | Microbiology Media |
| Packaging Type | 1 Ea. |
| Quantity | 100 mL |