Learn More
Thermo Scientific™ Remel™ BHI Broth (Brain Heart Infusion Broth)
Product Code. 17742998
Click to view available options
Quantity:
20 x 4 mL
20 x 5 mL
100 x 4 mL
100 x 5 mL
Format:
15mm x 103mm Tube
Unit Size:
Pack of 100
Pack of 20
Product Code. 17742998
Brand: Thermo Scientific™ R060264
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.
Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM, SMD, SMWW, USDA.
Description BHI Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 100 x 4 mL
REF R14101 for use with MicroScan™ System. REF R112022 not intended for IVD use.