Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Proteus O
Click to view available options
Description:
Agglutinating Sera, Proteus O, OX2 Positive Controls
Proteus OX19
Proteus OXK
Quantity:
2 mL
Detectable Analytes:
Proteus Suspension
Unit Size:
2mL
Description
Remel Agglutinating Sera, Proteus O are used as positive controls for stained Proteus suspensions.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Proteus OXK |
| CE Marker | Yes |
| Detectable Analytes | Proteus Suspension |
| Format | Vial |
| Packaging Type | 2 mL/Vial |
| Quantity | 2 mL |