Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis
Click to view available options
Description:
Group A
Group B
Group C
Group D
Group W135
Unit Size:
2mL
Description
Remel Agglutinating Sera are used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
Specifications
Specifications
| Description | Group B |
| CE Marker | Yes |
| Detectable Analytes | N. meningitidis |
| Type | Neisseria Meningitidis Test Kit |
| Organism Group | Group B |
| Quantity | Each |
| Target Organism Class | Neisseria Meningitidis |