missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RELN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20689-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Reelin in an extracellular matrix serine protease that plays a role in layering of neurons in the cerebral cortex and cerebellum. Regulates microtubule function in neurons and neuronal migration. Affects migration of sympathetic preganglionic neurons in the spinal cord, where it seems to act as a barrier to neuronal migration. Enzymatic activity is important for the modulation of cell adhesion. Binding to the extracellular domains of lipoprotein receptors VLDLR and LRP8/APOER2 induces tyrosine phosphorylation of DAB1 and modulation of TAU phosphorylation.Specifications
RELN | |
Polyclonal | |
Unconjugated | |
RELN | |
PRO1598, reelin, RELN, RL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19699, 5649 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P78509, Q60841 | |
RELN | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |