missing translation for 'onlineSavingsMsg'
Learn More

RelA/NFkB p65, p Ser536 Antibody, Novus Biologicals™

Product Code. p-7200772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
0.05 mL
Unit Size:
0.05mL
25µL
Product Code. Quantity unitSize
18061406 25 μL 25µL
18730184 0.05 mL 0.05mL
2 options
This item is not returnable. View return policy

Product Code. 18061406

Brand: Novus Biologicals NB1008208825UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 5 publications

RelA/NFkB p65 Polyclonal specifically detects RelA/NFkB p65 in Human, Mouse, Rat, Fish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RelA/NFkB p65 (p Ser536)
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100
Formulation PBS [pH 7.4] 150mM NaCl and 50% glycerol (without Mg2+ and Ca2+) with 0.02% Sodium Azide
Gene Accession No. Q04206
Gene Alias MGC131774, nf kb p65, NF kB p65 phospho, NFkB p65, NF-kB p65, NFkB p65 phospho, NF-kB p65 phospho, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, phospho NF kB p65, phospho NFkB p65, phospho NF-kB p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Gene Symbols RELA
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human NF-kB p65 around the phosphorylation site of serine 536 (F-S-SP-I-A).
Molecular Weight of Antigen 65 kDa
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5970
Test Specificity NF-kB p65 (specific to Phospho-Ser536) detects endogenous levels of NF-kB p65 only when phosphorylated at serine 536.
Target Species Human, Rat, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.