Learn More
BINDER™ Reinforced Shelf
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system
Brand:  BINDER™ 8012-1429
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 16454219
  • 6412.84 NOK / Each
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
Description
Description
  • Reinforced shelf with a load capacity of 45 kg
  • Designed to be positioned at bottom level
  • Includes an additional attachment for operation of shaking device, stirring device, or roller bottle system
Specifications
Specifications
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system
Reinforced Shelf
45 kg
BINDER Chambers KBW 400 Growth Chambers with Light
Increasing Incubator Capacity with Additional Attachment for Operation of Shaking Device, Stirring Device, or Roller Bottle System