Learn More
BINDER™ Reinforced Shelf
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system
Brand: BINDER™ 8012-1494
Additional Details : Weight : 0.00100kg
Description
- Reinforced shelf with a load capacity of 45 kg
- Designed to be positioned at bottom level
- Includes an additional attachment for operation of shaking device, stirring device, or roller bottle system
Specifications
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system | |
Reinforced Shelf | |
45 kg |
BINDER Chambers KBWF 720 Growth Chambers with Light and Humidity | |
Increasing Incubator Capacity with Additional Attachment for Operation of Shaking Device, Stirring Device, or Roller Bottle System |