Learn More
BINDER™ Reinforced Rack
Product Code. 15601826 Shop All BINDER Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. 15601826
Brand: BINDER™ 80120638
Only null left
Add to basket
Add to basket
Material: Stainless Steel; For Use With: KBF 240, KBF P 240, KBF LQC 240, KB 240, KB 400, KBW 240, KBW 400, KBWF 240, KMF 240, KBF-S 240
  • Manufactured from stainless steel
  • With one set of securing elements (4 pieces)
  • Maximum load of 50 kg or 70 kg
Specifications
Description Rack, reinforced - stainless steel, with fasteners (1 set of 4)
For Use With (Equipment) BINDER Chambers KBF 240, KBF P 240, KBF LQC 240, KB 240, KB 400, KBW 240, KBW 400, KBWF 240, KMF 240, KBF-S 240
Holds Max. 70 kg
Material Stainless Steel
Type Rack
Includes 1 set of 4 fasteners