Learn More
Brady™ Reinforced Polypropylene No Photography Prohibition Signs
Product Code. 11750184
Change view
Click to view available options
Unit Size:
Each
Product Code. unitSize
11750184 Each
1 options
Product Code. 11750184
Brand: Brady™ 222973
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Material Polypropylene
Quantity 1 Ea.
Description Reinforced Polypropylene
Diameter (Metric) 100 mm
For Use With (Application) Prohibition Signs Photographing prohibited
Thickness 0.369 mm
Temperature Range (Metric) -40°C to 120°C
Shape Round
Displayed Text No Photography Allowed
Product Type No Photographs Prohibition Sign
Color Black on White