Learn More
Brady™ Reinforced Polypropylene Fire Safety Signs
Product Code. 11740574
Click to view available options
Material:
Polypropylene
Unit Size:
Each
Product Code. 11740574
Brand: Brady™ 223981
Only null left
Specifications
Alternate Part Number PIC 414-148X148-B7527
Material Polypropylene
Quantity 1 Ea.
Description Reinforced Polypropylene
For Use With (Application) Used for a wide variety of applications both indoors and outdoors
Height (Metric) 148 mm
Thickness 0.369 mm
Width (Metric) 148 mm
Temperature Range (Metric) -40°C to 120°C
Shape Square
Displayed Text Fire blanket
Product Type Fire Safety Sign
Color White on Red