Learn More
Brady™ Reinforced Photoluminescent Aluminum Anti-Slip Strip
Click to view available options
Color:
Photoluminescent
Photoluminescent, Black
Dimensions (L x W):
0.80 m x 30 mm
0.80 m x 50 mm
Unit Size:
Each
Description
- Self-adhesive aluminium anti-skid strip is ideal for marking the edges of stairs and doorways.
- Available in different luminous intensities.
Specifications
Specifications
| Description | Photoluminescent Anti-skid Strips - Aluminium High intensity / Striped |
| Dimensions (L x W) | 0.80 m x 30 mm |
| For Use With (Application) | Exit and directional signs |
| Length (English) | 2.6 ft. |
| Length (Metric) | 0.8 m |
| Packaging Quantity | 1 Ea. |
| Temperature Range (English) | -40°F to +230°F |
| Temperature Range (Metric) | -40°C to +110°C |
| Thickness | 0.148 mm |
| Width (English) | 1.18 in. |
| Show More |