Description
- Reinforced inner chamber for bulky and heavy samples.
- Weight capacity ranged from 100 to 200 kg, depending on model.
Specifications
Specifications
| For Use With (Equipment) | KB PRO 1060 |
| Material | Stainless Steel |
| Type | Chamber Rack |
| Load Bearing Capacity (Metric) | 200 kg |