Learn More
Reichert™ Reichert™ Rhino Ultra-Durable Handheld Refractometers
Product Code. 10309714
Click to view available options
Accuracy:
±0.2
±0.25
Scale Range:
0 to 30 Brix
0 to 50 Brix
50 to 90 Brix
Increments:
0.2 and 0.5
0.2%
0.25%
Refractive Index:
1.333 to 1.3811
1.333 to 1.42
1.4201 to 1.5177
Unit Size:
Each
Product Code. 10309714
Brand: Reichert™ 137531L0
Only null left
Add to basket
Add to basket
Designed specifically for rugged factory and field use—fully IIP67 water rresistant, dustproof, and impact resistant
  • Automatic Temperature Compensation eliminates the need for temperature correction charts
  • Optics and instrument scales are permanently sealed inside the refractometer body
  • Magnified scales have easy-to-read markings and divisions for true readings and accurate results
  • Models are offered with Brix scales of 0-30, 0-50 and 50-90%
Order Info
Rhino Model for Industrial Fluid Testing is also available; see Cat. No. RE-10440.
Specifications
Type Handheld Ultra-durable Refractometer
Scale Range 0 to 50 Brix
Refractive Index 1.333 to 1.42
Accuracy ±0.25
Increments 0.25%
For Use With (Application) Food and beverage, chemical, pharmaceutical and industrial cutting fluids
Certifications/Compliance IIP67