Learn More
3M™ Regulator Assembly, S-212
Description
- Spare or replacement part
- Designed for use with 3M™ Versaflo™ Supplied Air System S-200+
- CE approved
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | 3M Versaflo Supplied Air System S-200+ |
| Product Type | SAR Parts and Accessories |