Learn More
REG Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15850-1-AP-20UL
 View more versions of this product
Product Code. 16855224
  • 1710.33 NOK / 20µL
Description
The REG antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human REG. This antibody recognizes human, mouse antigen. The REG antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
REG
Polyclonal
Unconjugated
20 μL
5967
IgG
Western Blot, Immunohistochemistry, ELISA
233 μg/mL
Rabbit
Primary
Human, Mouse