Learn More
Techne™ Refrigerated Bath
Click to view available options
Model:
RB-12A
RB-5A
Capacity (Metric):
7 L
12 L
Unit Size:
Each
Description
- Built-in refrigeration for efficient cooling
- Robust stainless steel construction makes for easy cleaning
Specifications
Specifications
| Capacity (Metric) | 7 L |
| Cooling Capacity | 20°C at 145W, 0°C at 145W, -10° C at 110W |
| Temperature Range | -20°C to +100°C |
| Features | Circulating bath with integrated refrigeration unit, Temperature range from -35°C to +100°C, Robust stainless-steel construction, Three models of refrigerated bath available: 7, 12, 22 liter,3 year warranty. |
| Model | RB-5A |
| Warranty | 3 Years |
| Voltage | 230 V |
| Frequency | 50/60 Hz |
Product Content Correction
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu