Learn More
Abeba™ Reflexor™ 36903 Shoes
Product Code. 15690798
Click to view available options
Size:
36
37
38
39
40
41
42
Unit Size:
1 pair
Product Code. 15690798
Brand: Abeba™ 36903/38
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ESD dissipative shoe includes PU-sole with high slip resistance.
  • CE EN ISO 20347:2012 OB A, E SRB
  • Smooth leather white
  • Slip resistance PU outsole sole
  • 2 replaceable Reflexor™ insoles (article no. 3565/ 3567)
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Instep area with elastic band
  • Buckle fastening
  • Hand-sewn seam
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRB
Color White
Liner Material Microfiber
Material Leather
Size 38
Sole Features Slip-resistant
Sole Material Polyurethane
Style 36903