Learn More
Tork Reflex Wiping Paper Plus M3
GREENER_CHOICE
Product Code. 30243794 Shop All Tork Products
Change view
Click to view available options
Color:
White
Unit Size:
9 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
30243794 White 9 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30243794 Supplier Tork Supplier No. 473474
  • ONLINE EXCLUSIVE PRICE
    1270.00 NOK
    1204.68 NOK / 9 rolls
    Save 65.32 NOK
    5% Off
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
The 2-ply multipurpose Tork Reflex Wiping Paper Plus is ideal for mopping up liquids and hand drying. This paper can be used in the Tork Mini Reflex single-sheet centerfeed dispenser, which fits your team’s workflow, cuts down consumption, and minimizes the risk of cross-contamination.
  • Cut down consumption by up to 37% with single-sheet dispensing, which makes it easy to complete one-handed tasks.
  • Improve hygiene with our Food Contact Approved roll - you touch only what you take.
  • No lengthy maintenance, thanks to SmartCore™ core removal - fast and easy refilling frees up time.
  • Avoid using multiple rolls with our multipurpose wiping paper that's strong and absorbent.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wiping Paper
Length (English) 73.27 yd.
Length (Metric) 67 m
Width (English) 7.64 in.
Width (Metric) 19.4 cm
Material Paper
Certifications/Compliance EU Ecolabel, FSC Chain-Of-Custody, BRC-IoP, ISO 9001, ISO 14001, ISO 45001, FSC Recycled credit
Color White
Packaging Quantity 9/Cs.
Ply 2-ply
Certifications/Compliance (Europe) EU Ecolabel, FSC Chain-Of-Custody, BRC-IoP, ISO 9001, ISO 14001, ISO 45001, FSC Recycled credit