Learn More
KGW Isotherm™ Refill for 2000mL Borosilicate Glass Cylindrical Dewar Container
Click to view available options
Capacity (Metric):
2 L
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Outer | 120mm |
| For Use With (Application) | Cooling of small samples, Storage: liquid nitrogen ( LN2 ), dry ice ( CO2 )other coolants |
| Height (Metric) Inner | 290mm |
| Height (Metric) | 345mm |
| Capacity (Metric) | 2 L |
| Diameter (Metric) Inner | 100 mm |
| Material | Borosilicate Glass |