Learn More
KGW Isotherm™ Refill for 1500mL Borosilicate Glass Cylindrical Dewar Container
Click to view available options
Capacity (Metric):
1500 mL
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Outer | 115mm |
| For Use With (Application) | Cooling of small samples, Storage: liquid nitrogen ( LN2 ), dry ice ( CO2 )other coolants |
| Height (Metric) Inner | 245mm |
| Height (Metric) | 300mm |
| Capacity (Metric) | 1500 mL |
| Diameter (Metric) Inner | 90 mm |
| Material | Borosilicate Glass |