missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
REEP6 may enhance the cell surface expression of odorant receptors.
Specifications
Specifications
| Antigen | REEP6 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.23 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | REEP6 |
| Gene Accession No. | Q5XI60, Q96HR9, Q9JM62 |
| Gene Alias | C19orf32, DP1L1, receptor accessory protein 6, REEP6, TB2L1 |
| Gene Symbols | Reep6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?