missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REEP6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12088-1-AP-20UL
This item is not returnable.
View return policy
Description
REEP6 may enhance the cell surface expression of odorant receptors.Specifications
| REEP6 | |
| Polyclonal | |
| Unconjugated | |
| REEP6 | |
| C19orf32, DP1L1, receptor accessory protein 6, REEP6, TB2L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362835, 70335, 92840 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5XI60, Q96HR9, Q9JM62 | |
| Reep6 | |
| REEP6 Fusion Protein Ag2723 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction