Learn More
BOLA Reducing Unions
Product Code. p-200110005
Click to view available options
Diameter (Metric) Outer:
8 mm
10 mm
12 mm
14 mm
16 mm
Thread Size:
4 mm
6 mm
8 mm
10 mm
Unit Size:
Each
Product Code. 10731611
Brand: BOLA D52604
Only null left
Add to basket
Add to basket
Reducing Unions PTFE, glass-fibre reinforced PTFE nut
Straight tube fitting made of PTFE with nuts made of glass-fibre reinforced PTFE. For connecting tubing or tube with different outer diameters. Universal chemical resistance, the product is only exposed to PTFE.
Specifications
Certifications/Compliance FDA
Autoclavable Autoclavable
Diameter (Metric) Outer 8 mm
Thread Size 4 mm
Color White