Learn More
Wheaton™ Red Replacement Cap for Urine Tube
Product Code. 16494959
Change view
Click to view available options
Closure Color:
Red
Unit Size:
Pack of 5000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color Unit Size
16494959 Red Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16494959 Supplier Wheaton™ Supplier No. 327226
  • ONLINE EXCLUSIVE PRICE
    4867.50 NOK
    3417.21 NOK / Pack of 5000
    Save 1450.29 NOK
    29% Off
Estimated Shipment: 12-01-2027
Spare polypropylene cap for primary urine tube.
  • Supports use of Wheaton™ primary urine tubes
  • For use with 10 mL tubes
Specifications
Product Type Cap
Closure Color Red
Material Polypropylene
Sterility Non-sterile
Description Replacement cap for primary urine tube with boric acid
For Use With (Equipment) 10 mL Urine Tube
No. per Case 5000/Cs.
No. per Pack 1000/Pk.
Storage Requirements Ambient