Learn More
Kautex™ Red Polypropylene Screw Closures
Product Code. 10548632
Change view
Click to view available options
For Use With (Equipment):
10 to 1000 mL Bottle
100 mL Bottle
1500/2500/4000 mL Bottle
250/350 mL Bottle
50 mL Bottle
500/750/1000 mL Bottle
Diameter (Metric):
28 mm
32 mm
45 mm
54 mm
80 mm
Unit Size:
Each
Pack of 100
Pack of 210
Product Code. For Use With (Equipment) Diameter (Metric) unitSize
10548632 500/750/1000 mL Bottle 54 mm Each
15941451 50 mL Bottle 28 mm Pack of 100
15901451 10 to 1000 mL Bottle 32 mm Pack of 100
10763281 100 mL Bottle 45 mm Each
10772761 1500/2500/4000 mL Bottle 80 mm Each
17828157 1500/2500/4000 mL Bottle 80 mm Pack of 210
10710851 250/350 mL Bottle 45 mm Each
7 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 10548632
Brand: Kautex™ 2000083691
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Seal a container's ingredients with these Kautex™ Red Polypropylene Screw Closures when you package, send samples, transport and store their contents.
  • PP
  • Red
  • Tamper evident ring
  • PE-foam insert
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Includes Tamper Evident Ring and Polyethylene Foam Insert
Material Polypropylene
Liner Material Polyethylene Foam Insert
Product Type Screw Cap
For Use With (Equipment) 500/750/1000 mL Bottle
Diameter (Metric) 54 mm