missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Red Screw Caps

Product Code. 11492340 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1000 Pk.
5000 Pk.
Unit Size:
Pack of 1000
Pack of 5000
Product Code. Quantity unitSize
11492340 5000 Pk. Pack of 5000
11482340 1000 Pk. Pack of 1000
2 options
This item is not returnable. View return policy

Product Code. 11492340

Brand: MP Biomedicals™ 115066010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use with empty FastPrep™ tubes. Red Caps are screw scaps with an O-ring for use with 2 mL microfuge tubes. These caps are sold separately for those who wish to fill their own Lysing Matrix C tubes for prepare samples for homogenization

  • Red Caps fit empty FastPrep™ tubes and are used in FastPrep systems.
  • Add sample to empty tube, add Lysing Matrix C material and lysing buffer, and apply cap. Process using FastPrep instrument or other bead-beater to lsye, grind or homogenize sample.
  • Red caps are also found in the Fast RNA™ Pro Red Kit for isolation of total RNA from yeast and fungi.
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Volume (Metric) 2 mL
Storage Requirements Store at Room Temperature (15°C to 30°C)
Quantity 5000 Pk.
Product Type Screw Cap

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.