Learn More
Gilac™ Rectangular Reinforced Stacking and Nesting Container
Product Code. 17988721
Change view
Click to view available options
Capacity (Metric):
9.6 L
40 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17988721 40 L Each
17978631 9.6 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17988721 Supplier Gilac™ Supplier No. G146421
  • ONLINE EXCLUSIVE PRICE
    470.40 NOK
    340.79 NOK / Pack of 1
    Save 129.61 NOK
    27% Off
Estimated Shipment: 21-10-2026
Rectangular Reinforced Stacking and Nesting Container is designed for intensive use and extreme handling conditions.
  • Reinforced containers for intensive use and extreme handling conditions.
  • Stackable and nestable by a quarter turn.
  • Highly durable.
  • Suitable for a number of different uses in the food processing, fishmonger, butchery, charcuterie, dairy and waste collection industries.
Specifications
Type Storage Tote
Material HDPE
Color White
Shape Rectangular
Capacity (English) 10.6 gal.
Capacity (Metric) 40 L