Learn More
Gilac™ Rectangular Container
Click to view available options
Volume (Metric):
12 L
15 L
25 L
35 L
55 L
Color:
Gray
White
Unit Size:
Each
Description
- Rectangular plastic food container.
- For sub-zero and ambient temperature storage and transporting raw materials.
- No gaps nor overlaps.
- Machine washable.
- Covering lid for increased hygiene, optional extra.
- Stackable without lid.
- Compatible with all GILAC 4 and 6-wheel dollies.
Specifications
Specifications
| Product Type | Food Container |
| Material | Polyethylene |
| Color | Gray |
| Shape | Rectangular |
| Volume (English) | 4 gal. |
| Volume (Metric) | 15 L |