Learn More
Helios Italquartz Rectangular Basins/Dishes, Transparent
Product Code. 15818044
Click to view available options
Dimensions (L x W):
60 x 12 mm
68 x 12 mm
115 x 12 mm
Unit Size:
Each
Product Code. 15818044
Brand: Helios Italquartz 62P00206
Only null left
Add to basket
Add to basket
Rectangular basins/dishes constructed of transparent, durable, resistant, and chemically inert quartz glass.
  • Transparent
  • Rectangular form
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Color Transparent
Material Quartz
Shape Rectangular
Type Dish
Dimensions (L x W) 115 x 12 mm
Length (Metric) 115/60 mm
Width (Metric) 12 mm
Height (Metric) 12 mm